Please login to your online account to display your discounted pricing

Mouse anti-CD15/Lewis X, Clone: HI98, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD15/Lewis X
Clone HI98
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD15.
Gene Accession No. P22083
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14367301

 
novus biologicals canada
NBP14367301MG
0.1mg Each for $252.00

NBP14367303

 
novus biologicals canada
NBP1436730025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CD15/Lewis X
Clone HI98
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD15.
Gene Accession No. P22083
Gene ID (Entrez) 2526
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgM, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)