Please login to your online account to display your discounted pricing

Mouse anti-CD11c, Clone: HC1/1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462901

  • / Each
 

Description & Specifications

Specifications

Antigen CD11c
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone HC1/1
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P20702
Host Species Murine
Immunogen Human monocytic U937 phorbol ester-treated cells
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3687
Reconstitution Protein A purified
Target Species Human