Please login to your online account to display your discounted pricing

Mouse anti-CD11c, Clone: HC1/1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462902

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Quantity 0.2mg
Conjugate Unlabeled
Host Species Mouse
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Gene Accession No. P20702
Format Purified
Concentration 1mg/mL
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 3687
Immunogen Human monocytic U937 phorbol ester-treated cells
Antigen CD11c
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Clone HC1/1
Primary or Secondary Primary
Purification Method Purified