missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CD117/c-kit, Clone: YB5.B8, Novus Biologicals™

$252.00 - $329.00

Specifications

Antigen CD117/c-kit
Clone YB5.B8
Concentration 0.5mg/mL
Immunogen Human CD117
Gene ID (Entrez) 3815
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14335801 Novus Biologicals Canada
NBP14335801MG
0.1mg Each for $329.00
NBP14335803 Novus Biologicals Canada
NBP1433580025MG
0.025mg Each for $252.00
Specifications Description & Specifications

Specifications

CD117/c-kit
0.5mg/mL
3815
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Affinity Purified
Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen),Immunoprecipitation
YB5.B8
Human CD117
Phosphate buffer pH 7.2
Primary
Murine
Unlabeled
Affinity Purified