Please login to your online account to display your discounted pricing

Mouse anti-CD117/c-kit, Clone: YB5.B8, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD117/c-kit
Clone YB5.B8
Concentration 0.5mg/mL
Immunogen Human CD117
Gene ID (Entrez) 3815
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14358901

 
novus biologicals canada
NBP14358901MG
0.1mg Each for $252.00

NBP14358903

 
novus biologicals canada
NBP1435890025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen CD117/c-kit
Clone YB5.B8
Concentration 0.5mg/mL
Immunogen Human CD117
Gene ID (Entrez) 3815
Formulation Phosphate buffered saline, pH 7.2 less than or equal to 0.09% NaN3
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry