Please login to your online account to display your discounted pricing

Mouse anti-CCR9, Clone: CW-1.2, PE, Novus Biologicals™

$171.00 - $354.00

Specifications

Antigen CCR9
Clone CW-1.2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD199.
Gene ID (Entrez) 10803
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14379301

 
novus biologicals canada
NBP14379301MG
0.1mg Each for $354.00

NBP14379303

 
novus biologicals canada
NBP1437930025MG
0.025mg Each for $171.00
Description & Specifications

Specifications

Antigen CCR9
Clone CW-1.2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD199.
Gene ID (Entrez) 10803
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry