Please login to your online account to display your discounted pricing

Mouse anti-CCR9, Clone: CW-1.2, Novus Biologicals™

$321.00 - $418.00

Specifications

Antigen CCR9
Clone CW-1.2
Immunogen The immunogen for this antibody was CD199.
Gene ID (Entrez) 10803
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14333101

 
novus biologicals canada
NBP14333101MG
0.1mg Each for $321.00

NBP1433315

 
novus biologicals canada
NBP14333105MG
0.5mg Each for $418.00
Description & Specifications

Specifications

Antigen CCR9
Clone CW-1.2
Immunogen The immunogen for this antibody was CD199.
Gene ID (Entrez) 10803
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG2a
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry