Please login to your online account to display your discounted pricing

Mouse anti-CCR9, Clone: CW-1.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14333101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14333101

  • / Each
 

Description & Specifications

Specifications

Antigen CCR9
Applications Flow Cytometry
Clone CW-1.2
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Immunogen The immunogen for this antibody was CD199.
Isotype IgG2a
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10803