Please login to your online account to display your discounted pricing

Mouse anti-CCR5, Clone: T21/8, PE, Novus Biologicals™

$131.00 - $362.00

Specifications

Antigen CCR5
Clone T21/8
Immunogen The immunogen for this antibody was CCR5.
Gene Accession No. P51681
Gene ID (Entrez) 1234
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200325

 
novus biologicals canada
NBP200325100TESTS
100 Tests Each for $362.00

NBP20032525

 
novus biologicals canada
NBP20032525TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CCR5
Clone T21/8
Immunogen The immunogen for this antibody was CCR5.
Gene Accession No. P51681
Gene ID (Entrez) 1234
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry