Please login to your online account to display your discounted pricing

Mouse anti-CCR5, Clone: T21/8, Novus Biologicals™

$274.00 - $329.00

Specifications

Antigen CCR5
Clone T21/8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCR5.
Gene Accession No. P51681
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14333501

 
novus biologicals canada
NBP14333501MG
0.1mg Each for $329.00

NBP14333503

 
novus biologicals canada
NBP1433350025MG
0.025mg Each for $274.00
Description & Specifications

Specifications

Antigen CCR5
Clone T21/8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCR5.
Gene Accession No. P51681
Gene ID (Entrez) 1234
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot