Please login to your online account to display your discounted pricing

Mouse anti-CCR5, Clone: T21/8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14333501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14333501

  • / Each
 

Description & Specifications

Specifications

Antigen CCR5
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone T21/8
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Gene Accession No. P51681
Host Species Murine
Immunogen The immunogen for this antibody was CCR5.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1234
Target Species Human