Please login to your online account to display your discounted pricing

Mouse anti-CCR5, Clone: T21/8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433350025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14333503

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 1234
Storage Requirements Store at 4°C. Do not freeze.
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen CCR5
Quantity 0.025mg
Isotype IgG1, κ
Concentration 0.5mg/mL
Conjugate Unlabeled
Host Species Mouse
Primary or Secondary Primary
Formulation PBS [pH 7.2]
Immunogen The immunogen for this antibody was CCR5.
Purification Method Affinity Purified
Regulatory Status RUO
Clone T21/8
Gene Accession No. P51681