Please login to your online account to display your discounted pricing

Mouse anti-CCR5, Clone: T21/8, Biotin, Novus Biologicals™

$128.00 - $289.00

Specifications

Antigen CCR5
Clone T21/8
Immunogen The immunogen for this antibody was CCR5.
Gene Accession No. P51681
Gene ID (Entrez) 1234
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14357201

 
novus biologicals canada
NBP14357201MG
0.1mg Each for $289.00

NBP14357203

 
novus biologicals canada
NBP1435720025MG
0.025mg Each for $128.00
Description & Specifications

Specifications

Antigen CCR5
Clone T21/8
Immunogen The immunogen for this antibody was CCR5.
Gene Accession No. P51681
Gene ID (Entrez) 1234
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)