Please login to your online account to display your discounted pricing

Mouse anti-CCR5, Clone: T21/8, Biotin, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP1435720025MG

 View all options for this product


For Research Use Only

Catalog No. NBP14357203


Description & Specifications


Antigen CCR5
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone T21/8
Conjugate Biotin
Cross Reactivity Human
Format Affinity Purified
Formulation PBS [pH 7.2]
Gene Accession No. P51681
Host Species Mouse
Immunogen The immunogen for this antibody was CCR5.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1234