Please login to your online account to display your discounted pricing

Mouse anti-CCR5, Clone: T21/8, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14357201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14357201

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen CCR5
Formulation PBS [pH 7.2]
Quantity 0.1mg
Regulatory Status RUO
Conjugate Biotin
Gene ID (Entrez) 1234
Clone T21/8
Immunogen The immunogen for this antibody was CCR5.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Host Species Mouse
Storage Requirements Store at 4°C. Do not freeze.
Format Affinity Purified
Gene Accession No. P51681
Isotype IgG1, κ
Purification Method Affinity Purified
Cross Reactivity Human