Please login to your online account to display your discounted pricing

Mouse anti-CCR3, Clone: 5E8-G9-B4, PE, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CCR3
Clone 5E8-G9-B4
Immunogen The immunogen for this antibody was CCR3.
Gene ID (Entrez) 1232
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143799

 
novus biologicals canada
NBP143799100TESTS
100 Tests Each for $252.00

NBP14379925

 
novus biologicals canada
NBP14379925TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen CCR3
Clone 5E8-G9-B4
Immunogen The immunogen for this antibody was CCR3.
Gene ID (Entrez) 1232
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry