Please login to your online account to display your discounted pricing

Mouse anti-CCR3, Clone: 5E8-G9-B4, Novus Biologicals™

$274.00 - $329.00

Specifications

Antigen CCR3
Clone 5E8-G9-B4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCR3.
Gene ID (Entrez) 1232
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14333701

 
novus biologicals canada
NBP14333701MG
0.1mg Each for $329.00

NBP14333703

 
novus biologicals canada
NBP1433370025MG
0.025mg Each for $274.00
Description & Specifications

Specifications

Antigen CCR3
Clone 5E8-G9-B4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CCR3.
Gene ID (Entrez) 1232
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry