Please login to your online account to display your discounted pricing

Mouse anti-CCR3, Clone: 5E8-G9-B4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433370025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14333703

  • / Each
 

Description & Specifications

Specifications

Antigen CCR3
Applications Flow Cytometry
Clone 5E8-G9-B4
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Murine
Immunogen The immunogen for this antibody was CCR3.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1232
Target Species Human