Please login to your online account to display your discounted pricing

Mouse anti-CCR3, Clone: 5E8-G9-B4, APC, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen CCR3
Clone 5E8-G9-B4
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CCR3.
Gene ID (Entrez) 1232
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143013

 
novus biologicals canada
NBP143013100TESTS
100 Tests Each for $362.00

NBP14301325

 
novus biologicals canada
NBP14301325TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen CCR3
Clone 5E8-G9-B4
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CCR3.
Gene ID (Entrez) 1232
Formulation PBS [pH 7.2] containing BSA and/or gelatin for protein stabilization
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG2b, κ
Conjugate Allophycocyanin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry