Please login to your online account to display your discounted pricing

Mouse anti-CCL5/RANTES, Clone: R6G9, Novus Biologicals™

$246.00 - $300.00

Specifications

Antigen CCL5/RANTES
Clone R6G9
Immunogen The immunogen for this antibody was CCL5.
Gene ID (Entrez) 6352
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14324401

 
novus biologicals canada
NBP14324401MG
0.1mg Each for $300.00

NBP14324403

 
novus biologicals canada
NBP1432440025MG
0.025mg Each for $246.00
Description & Specifications

Specifications

Antigen CCL5/RANTES
Clone R6G9
Immunogen The immunogen for this antibody was CCL5.
Gene ID (Entrez) 6352
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Western Blotting