Please login to your online account to display your discounted pricing

Mouse anti-CCL5/RANTES, Clone: R6G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1432440025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14324403

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6352
Antigen CCL5/RANTES
Formulation PBS [pH 7.2]
Immunogen The immunogen for this antibody was CCL5.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Clone R6G9
Cross Reactivity Mouse
Format Affinity Purified
Isotype IgG1
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled