Please login to your online account to display your discounted pricing

Mouse anti-CCL5/RANTES, Clone: R6G9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14324401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14324401

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Formulation PBS [pH 7.2]
Quantity 0.1mg
Primary or Secondary Primary
Cross Reactivity Mouse
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Immunogen The immunogen for this antibody was CCL5.
Antigen CCL5/RANTES
Purification Method Affinity Purified
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Clone R6G9
Format Affinity Purified
Gene ID (Entrez) 6352
Conjugate Unlabeled