Please login to your online account to display your discounted pricing

Mouse anti-CCL16/HCC-4/LEC, Clone: LEC67, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285801

  • / Each
 

Description & Specifications

Specifications

Antigen CCL16/HCC-4/LEC
Format Purified
Formulation PBS with 0.05% BSA.
Purification Method Purified
Quantity 0.1mL
Gene ID (Entrez) 6360
Reconstitution Protein A purified
Clone LEC67
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. O15467
Host Species Mouse
Applications ELISA
Immunogen Recombinant human HCC-4 protein
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human