Please login to your online account to display your discounted pricing

Mouse anti-CCL16/HCC-4/LEC, Clone: LEC67, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466701

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Isotype IgG1, κ
Host Species Mouse
Regulatory Status RUO
Cross Reactivity Human
Conjugate Unlabeled
Applications ELISA
Applications Functional Assay
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Purification Method Purified
Antigen CCL16/HCC-4/LEC
Immunogen Recombinant human HCC-4 protein
Monoclonal or Polyclonal Monoclonal
Format Purified
Reconstitution Protein A purified
Concentration 1mg/mL
Gene ID (Entrez) 6360
Clone LEC67
Gene Accession No. O15467
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.