Please login to your online account to display your discounted pricing

Mouse anti-CCL16/HCC-4/LEC, Clone: LEC66, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466601

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Functional Assay
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 6360
Format Purified
Conjugate Unlabeled
Purification Method Purified
Reconstitution Protein A purified
Clone LEC66
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Antigen CCL16/HCC-4/LEC
Gene Accession No. O15467
Immunogen Recombinant human HCC-4 protein
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Concentration 1mg/mL
Cross Reactivity Human