missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Cathepsin Z, Clone: 6G11, Novus Biologicals™

$254.34 - $310.66

Specifications

Antigen Cathepsin Z
Clone 6G11
Concentration 1mg/mL
Immunogen Recombinant human CTSZ (62-303aa) purified from E. coli
Gene Accession No. NP_001327
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23121101 Novus Biologicals Canada
NBP23121101ML
0.1mL Each for $310.66
NBP23121105 Novus Biologicals Canada
NBP231211005ML
0.05mL Each for $254.34
Specifications Description & Specifications

Specifications

Cathepsin Z
1mg/mL
NP_001327
Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.02%
Primary
Murine
Unlabeled
Purified
6G11
Recombinant human CTSZ (62-303aa) purified from E. coli
1522
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
ELISA,Western Blot