Please login to your online account to display your discounted pricing

Mouse anti-Cathepsin Z, Clone: 6G11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231211005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23121105

  • / Each
 

Description & Specifications

Specifications

Format Purified
Conjugate Unlabeled
Quantity 0.05mL
Applications ELISA
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Purification Method Purified
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol, 0.02%
Reconstitution Protein A purified
Cross Reactivity Human
Immunogen Recombinant human CTSZ (62-303aa) purified from E. coli
Isotype IgG2b, κ
Antigen Cathepsin Z
Gene ID (Entrez) 1522
Host Species Mouse
Gene Accession No. NP_001327
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone 6G11
Concentration 1mg/mL
Primary or Secondary Primary