Please login to your online account to display your discounted pricing

Mouse anti-Carbonic Anhydrase IX/CA9, Clone: SPM487, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23436301ML

 View all options for this product


For Research Use Only

Catalog No. NBP23436301


Description & Specifications


Antigen Carbonic Anhydrase IX/CA9
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM487
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16790
Host Species Mouse
Immunogen Microsomal fraction of human renal cortical tissue homogenate
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 768
Test Specificity Recognizes a glycoprotein of ~200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200.
Reconstitution Protein A purified