Please login to your online account to display your discounted pricing

Mouse anti-Carbonic Anhydrase IX/CA9, Clone: SPM487, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441101

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Equine
Cross Reactivity Human
Antigen Carbonic Anhydrase IX/CA9
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q16790
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Microsomal fraction of human renal cortical tissue homogenate
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 768
Test Specificity Recognizes a glycoprotein of ~200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200.
Reconstitution Protein A purified
Clone SPM487
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL