Please login to your online account to display your discounted pricing

Mouse anti-Carbonic Anhydrase IX/CA9, Clone: SPM314, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Carbonic Anhydrase IX/CA9
Clone SPM314
Concentration 0.2mg/mL
Immunogen Microsomal fraction of human renal cortical tissue homogenate
Gene Accession No. Q16790
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23436001

 
novus biologicals canada
NBP23436001ML
0.1mL Each for $139.00

NBP23436005

 
novus biologicals canada
NBP23436005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Carbonic Anhydrase IX/CA9
Clone SPM314
Concentration 0.2mg/mL
Immunogen Microsomal fraction of human renal cortical tissue homogenate
Gene Accession No. Q16790
Gene ID (Entrez) 768
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Equine
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting