Please login to your online account to display your discounted pricing

Mouse anti-Calreticulin, Clone: FMC 75, Novus Biologicals™

$310.66 - $582.00

Specifications

Antigen Calreticulin
Clone FMC 75
Concentration 1.0mg/mL
Immunogen Recombinan Calreticulin-maltose binding fusion protein.
Gene Accession No. P27797
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19750202

 
novus biologicals canada
NBP19750202MG
0.2mg Each for $582.00

NBP19750205

 
novus biologicals canada
NBP197502005MG
0.05mg Each for $310.66
Description & Specifications

Specifications

Antigen Calreticulin
Clone FMC 75
Concentration 1.0mg/mL
Immunogen Recombinan Calreticulin-maltose binding fusion protein.
Gene Accession No. P27797
Gene ID (Entrez) 811
Formulation PBS and 50% Glycerol
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Primate
Cross Reactivity Recognizes Primate Calreticulin.
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Protein G purified
Applications Immunocytochemistry/Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting