Please login to your online account to display your discounted pricing

Mouse anti-Calreticulin, Clone: FMC 75, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19750202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19750202

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry/Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Protein G purified
Cross Reactivity Primate
Cross Reactivity Recognizes Primate Calreticulin.
Gene ID (Entrez) 811
Formulation PBS and 50% Glycerol
Species Reactivity Human, Primate
Gene Alias autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP
Immunogen Recombinan Calreticulin-maltose binding fusion protein.
Primary or Secondary Primary
Host Species Mouse
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Antigen Calreticulin
Quantity 0.2mg
Isotype IgG1
Conjugate Unlabeled
Gene Accession No. P27797
Test Specificity Recognizes Primate Calreticulin.
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Clone FMC 75
Format Purified

Calreticulin Monoclonal antibody specifically detects Antigen in Human, Primate samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.