Please login to your online account to display your discounted pricing

Mouse anti-Calnexin, Clone: IE2.1C12, Novus Biologicals™

$109.00 - $307.00

Specifications

Antigen Calnexin
Clone IE2.1C12
Concentration 1.0mg/mL
Immunogen Partial recombinant human Calnexin protein (between amino acids 1-300) [UniProt P27824]
Gene Accession No. P27824
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236571SS

 
novus biologicals canada
NBP236571SS
0.025mg Each for $109.00

NBP236571

 
novus biologicals canada
NBP236571
0.1mg Each for $307.00
Description & Specifications

Specifications

Antigen Calnexin
Clone IE2.1C12
Concentration 1.0mg/mL
Immunogen Partial recombinant human Calnexin protein (between amino acids 1-300) [UniProt P27824]
Gene Accession No. P27824
Gene ID (Entrez) 821
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow (Intracellular)
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting