Please login to your online account to display your discounted pricing

Mouse anti-CA19-9/Sialyl Lewisa, Clone: SPM588, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CA19-9/Sialyl Lewisa
Clone SPM588
Immunogen Precipitin lines obtained after immuno-diffusion using MAb 116-NS-19-9 and mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le).
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23445805

 
novus biologicals canada
NBP23445805ML
0.5mL Each for $321.00

NBP23445801

 
novus biologicals canada
NBP23445801ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen CA19-9/Sialyl Lewisa
Clone SPM588
Immunogen Precipitin lines obtained after immuno-diffusion using MAb 116-NS-19-9 and mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le).
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting