Please login to your online account to display your discounted pricing

Mouse anti-CA19-9/Sialyl Lewisa, Clone: SPM588, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445805

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Precipitin lines obtained after immuno-diffusion using MAb 116-NS-19-9 and mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le).
Conjugate Unlabeled
Cross Reactivity Human
Quantity 0.5mL
Host Species Mouse
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Isotype IgM, κ
Antigen CA19-9/Sialyl Lewisa
Clone SPM588
Regulatory Status RUO
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Concentrated
Format Concentrated