Please login to your online account to display your discounted pricing

Mouse anti-CA19-9, Clone: 121SLE, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CA19-9
Clone 121SLE
Immunogen Precipitin lines obtained after immunodiffusion using MAb 116-NS-19-9 and mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le) were used as immunogen for this CA19-9 antibody.
Gene Accession No. P50406
Gene ID (Entrez) 3362
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21518201

 
novus biologicals canada
NBP21518201ML
0.1mL Each for $139.00

NBP21518210

 
novus biologicals canada
NBP21518210ML
1mL Each for $429.00

NBP21518205

 
novus biologicals canada
NBP21518205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CA19-9
Clone 121SLE
Immunogen Precipitin lines obtained after immunodiffusion using MAb 116-NS-19-9 and mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le) were used as immunogen for this CA19-9 antibody.
Gene Accession No. P50406
Gene ID (Entrez) 3362
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)