Please login to your online account to display your discounted pricing

Mouse anti-CA19-9, Clone: 121SLE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21518201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21518201

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Antigen CA19-9
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P50406
Immunogen Precipitin lines obtained after immunodiffusion using MAb 116-NS-19-9 and mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le) were used as immunogen for this CA19-9 antibody.
Isotype IgM, κ
Quantity 0.1mL
Gene ID (Entrez) 3362
Purification Method Concentrated
Test Specificity CA19-9, a carbohydrate epitope expressed on a high MW (>400kDa) mucin glycoprotein, is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas, including adenocarcinomas of the stomach, intestine, and pancreas. Preoperative elevated CA19-9 levels in patients with stage I pancreatic carcinoma decrease to normal values following surgery. When used serially, CA19-9 can predict recurrence of disease prior to radiographic or clinical findings. This MAb is superb for staining of formalin-fixed, paraffin-embedded tissues.
Clone 121SLE
Primary or Secondary Primary