Please login to your online account to display your discounted pricing

Mouse anti-C1q R1/CD93, Clone: R3, Biotin, Novus Biologicals™

$131.00 - $329.00

Specifications

Antigen C1q R1/CD93
Clone R3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD93.
Gene ID (Entrez) 22918
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14360501

 
novus biologicals canada
NBP14360501MG
0.1mg Each for $329.00

NBP14360503

 
novus biologicals canada
NBP1436050025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen C1q R1/CD93
Clone R3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD93.
Gene ID (Entrez) 22918
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgM
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Flow Cytometry