Please login to your online account to display your discounted pricing

Mouse anti-c-Myc, Clone: MYC699, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469201

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Chimpanzee
Cross Reactivity Gorilla
Cross Reactivity Human
Cross Reactivity Primate
Antigen c-Myc
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4609
Conjugate Unlabeled
Gene Accession No. P01106
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Mouse (Negative)
Reconstitution Protein A purified
Clone MYC699
Concentration 1mg/mL