Please login to your online account to display your discounted pricing

Mouse anti-c-Myc, Clone: MYC275, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen c-Myc
Clone MYC275
Concentration 1mg/mL
Immunogen Recombinant human c-myc protein
Gene Accession No. P01106
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23469101

 
novus biologicals canada
NBP23469101MG
0.1mg Each for $384.00

NBP23469102

 
novus biologicals canada
NBP23469102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen c-Myc
Clone MYC275
Concentration 1mg/mL
Immunogen Recombinant human c-myc protein
Gene Accession No. P01106
Gene ID (Entrez) 4609
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot