Please login to your online account to display your discounted pricing

Mouse anti-c-Myc, Clone: MYC275, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469102

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4609
Primary or Secondary Primary
Regulatory Status RUO
Conjugate Unlabeled
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Quantity 0.2mg
Antigen c-Myc
Host Species Mouse
Format Purified
Clone MYC275
Gene Accession No. P01106
Immunogen Recombinant human c-myc protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Concentration 1mg/mL
Cross Reactivity Human