Please login to your online account to display your discounted pricing

Mouse anti-c-Myc, Clone: MYC275, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469101

  • / Each
 

Description & Specifications

Specifications

Antigen c-Myc
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Recombinant human c-myc protein
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4609
Conjugate Unlabeled
Gene Accession No. P01106
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone MYC275
Concentration 1mg/mL
Cross Reactivity Human