Please login to your online account to display your discounted pricing

Mouse anti-c-Myc, Clone: CT14.G4, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460001

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Chimpanzee
Cross Reactivity Gorilla
Cross Reactivity Human
Cross Reactivity Primate
Antigen c-Myc
Format Purified
Gene Accession No. P01106
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity Mouse (Negative)
Reconstitution Protein A purified
Clone CT14.G4
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.1mg
Gene ID (Entrez) 4609