Please login to your online account to display your discounted pricing

Mouse anti-c-Myb, Clone: MYB286, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Antigen c-Myb
Regulatory Status RUO
Conjugate Unlabeled
Immunogen A synthetic peptide
Gene ID (Entrez) 4602
Gene Accession No. P10242
Purification Method Purified
Clone MYB286
Concentration 1mg/mL
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1, κ
Reconstitution Protein A purified
Format Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal