Please login to your online account to display your discounted pricing

Mouse anti-Bromodeoxyuridine/BrdU, Clone: SPM537, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23292110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23292110

  • / Each
 

Description & Specifications

Specifications

Antigen Bromodeoxyuridine/BrdU
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Isotype IgG1
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM537
Conjugate Unlabeled
Cross Reactivity All
Concentration 0.2mg/mL