Please login to your online account to display your discounted pricing

Mouse anti-Bromodeoxyuridine/BrdU, Clone: SPM537, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity All
Conjugate Unlabeled
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Antigen Bromodeoxyuridine/BrdU
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Format Purified
Host Species Mouse
Isotype IgG1
Test Specificity Expected to show broad species reactivity.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Clone SPM537
Purification Method Purified
Reconstitution Protein A purified
Quantity 0.1mg
Primary or Secondary Primary