Please login to your online account to display your discounted pricing

Mouse anti-Bromodeoxyuridine/BrdU, Clone: SPM166, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23291901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23291901

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity All
Antigen Bromodeoxyuridine/BrdU
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Isotype IgG1
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM166
Concentration 0.2mg/mL
Conjugate Unlabeled
Host Species Mouse