missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Bromodeoxyuridine/BrdU, Clone: SPM166, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Bromodeoxyuridine/BrdU
Clone SPM166
Concentration 1mg/mL
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23474401 Novus Biologicals Canada
NBP23474401MG
0.1mg Each for $384.00
NBP23474402 Novus Biologicals Canada
NBP23474402MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Bromodeoxyuridine/BrdU
1mg/mL
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM166
Bromodeoxyuridine (BrdU) conjugated to KLH
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)