Please login to your online account to display your discounted pricing

Mouse anti-Bromodeoxyuridine/BrdU, Clone: SPM166, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474402

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity All
Host Species Mouse
Conjugate Unlabeled
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen Bromodeoxyuridine/BrdU
Concentration 1mg/mL
Reconstitution Protein A purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.2mg
Purification Method Purified
Format Purified
Isotype IgG1
Clone SPM166
Monoclonal or Polyclonal Monoclonal
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Regulatory Status RUO
Test Specificity It reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.