missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Bromodeoxyuridine/BrdU, Clone: BRD494, Azide Free, Novus Biologicals™

$335.73 - $515.00

Specifications

Antigen Bromodeoxyuridine/BrdU
Clone BRD494
Concentration 1mg/mL
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23456602 Novus Biologicals Canada
NBP23456602MG
0.2mg Each for $515.00
NBP23456601 Novus Biologicals Canada
NBP23456601MG
0.1mg Each for $335.73
Specifications Description & Specifications

Specifications

Bromodeoxyuridine/BrdU
1mg/mL
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
BRD494
Bromodeoxyuridine (BrdU) conjugated to KLH
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)